missing translation for 'onlineSavingsMsg'
Learn More

Silver trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

£27.34 - £277.38

Chemical Identifiers

CAS 2923-28-6
Molecular Formula CAgF3O3S
Molecular Weight (g/mol) 256.93
MDL Number MFCD00013226
InChI Key QRUBYZBWAOOHSV-UHFFFAOYSA-M
Synonym silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
PubChem CID 76223
IUPAC Name silver;trifluoromethanesulfonate
SMILES [Ag+].[O-]S(=O)(=O)C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389339
View Documents
Thermo Scientific Alfa Aesar
088722.04
2 g
£27.34
2g
Please to purchase this item. Need a web account? Register with us today!
11379339
View Documents
Thermo Scientific Alfa Aesar
088722.09
10 g
£73.06
10g
Please to purchase this item. Need a web account? Register with us today!
11369339
View Documents
Thermo Scientific Alfa Aesar
088722.18
50 g
£277.38
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Silver trifluoromethanesulfonate is useful in halide abstraction. It helps in activating sulphur protecting groups at the anomeric centres of carbohydrates in the presence of iodine. It is employed as a triflating agent in combination with Palladium compounds, as a catalyst for cross coupling reactions, and in several organic transformation reactions. Polyurethanes prepared from silver triflate exhibit high permeability and selectivity towards olefins in their separation from paraffin mixtures. Polyethylene oxide - silver triflate composites find application in humidity sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2923-28-6
256.93
QRUBYZBWAOOHSV-UHFFFAOYSA-M
76223
[Ag+].[O-]S(=O)(=O)C(F)(F)F
CAgF3O3S
MFCD00013226
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
silver;trifluoromethanesulfonate

Specifications

356°C
White
98%
MFCD00013226
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
QRUBYZBWAOOHSV-UHFFFAOYSA-M
silver;trifluoromethanesulfonate
76223
Odorless
Silver trifluoromethanesulfonate
2923-28-6
Crystalline
CAgF3O3S
EM>10,
Soluble in water and also in most organic solvents.
[Ag+].[O-]S(=O)(=O)C(F)(F)F
256.93
256.93
Light sensitive
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III

missing translation for 'einecsNumber' : 220-882-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Silver trifluoromethanesulfonate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Silver trifluoromethanesulfonate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only