missing translation for 'onlineSavingsMsg'
Learn More

Stomacher™ Simplettes

The Simplettes™ provide an accurate and economical alternative to pipette tips. They are designed specifically to pipette samples from the bottom of the Stomacher™ bag without risk of cross contamination.

Brand:  Stomacher™ BA6053

 View more versions of this product

Product Code. 17784399

  • £45.70 / Pack of 2000
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Single use pipetting straws
  • Capable of pipetting volumes between 0.1 and 2 mL
  • Irradiated sterile
  • Available in various pack sizes
Specifications

Specifications

Tip
3.6 to 3.7 mm (Internal diameter)
177 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.