Learn More
Socorex™ Acura™ Single-channel Macropipette Volumetric Module
Increase the versatility of your Acura™ electronic macropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ Acura™ Single-channel Macropipette Volumetric Module is an excellent and easily attachable/detachable choice for your single-channel macropipetting needs.
£87.95
Specifications
For Use With Acura™ Electro™ 926 MicroPipette
Number of Channels 1
Product Line Acura
Type Electronic Pipette Volumetric Module
For Use With (Equipment) Acura™ Electro 926 Micropipette
View More Specs
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10265282
View Documents
Socorex™
800.5000
0.25 to 5 mL
£87.95
Each
Estimated Shipment: 02-07-2025
to see stock.
Add to basket
10632203
View Documents
Socorex™
800.10000
0.5 to 10 mL
£87.95
Each
Estimated Shipment: 18-06-2025
to see stock.
Add to basket
Description
Description
  • One of 27 available volumetric assemblies.
    • Increase versatility and save costs.
  • Disassembles and reassembles without tools.
  • Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
Specifications
Acura™ Electro™ 926 MicroPipette
Acura
Acura™ Electro 926 Micropipette
1
Electronic Pipette Volumetric Module
1 Each
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Socorex™ Acura™ Single-channel Macropipette Volumetric Module