Learn More
Gilson™ Single Channel PIPETMAN™ M
GREENER_CHOICE
Single Channel PIPETMAN™ M is a Bluetooth™-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes.
£621.00 - £694.00
Specifications
Compatible Tips Universal
Adjustable Tip Spacing No
Autoclavable Autoclavable
For Use With Pipette Tip, TRACKMAN™ Connected
Material Polypropylene
View More Specs
Products 8
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
16348147
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F81040
0.5 to 10 μL
£638.00
Each
16358147
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F81041
2 to 20 μL
£621.00
Each
16368147
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F81042
5 to 100 μL
£694.00
Each
16378147
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F81043
20 to 200 μL
£621.00
Each
16388147
GREENER_CHOICE
View Documents
Gilson™
F81044
20 to 300 μL
£621.00
Each
16398147
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F81045
100 to 1200 μL
£622.00
Each
16308157
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F81046
500 to 5000 μL
£622.00
Each
16318157
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F81047
1 to 10 mL
£622.00
Each
Description
Description
  • Allows accurate and precise pipetting
  • The motorized piston allows for high pipetting reproducibility and accuracy in all modes
  • Associated with TRACKMAN™ Connected and its PipettePilot™ application, PIPETMAN™ M Connected reduces the risk of human error by transferring verifiable pipetting parameters and commands in the pipette
  • Increased functionality
  • The intuitive interface has five pipetting modes for a large number of applications: pipette, repetitive, mix, reverse, and custom
  • Create and transfer up to 10 custom protocols from PIPETMAN™ M Connected Software into your pipette
  • Bluetooth™ connection enables communication with TRACKMAN™ Connected and helps guide you through your protocol telling you where, when, and how much to pipette with real-time interactions
  • Simple and comfortable to use
  • Aspirate and dispense with one click of the push button and eject tips with ease
  • The lightweight and ergonomic design rests comfortably in your hand for maximum pipetting comfort
Specifications
Specifications
Universal
Autoclavable
Polypropylene
Pipetman M
Non-sterile
Pipette Tips, TRACKMAN™ Connected
12.75 in.
8 in.
6.4 cm
Certified Quality
No
Pipette Tip, TRACKMAN™ Connected
1
Yes
Pipette
1 Year
32.4 cm
2.5 in.
3.7 V and 420 mAh (1.55 Wh)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Gilson™ Single Channel PIPETMAN™ M