missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Single-Use Filler Nozzles

Reduce filler line downtime and risk of cross-contamination.

Brand:  Cole-Parmer™ 30619-07

 View more versions of this product

Product Code. 12677107

  • £322.00 / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Cole-Parmer Nozzle Filler Fittings For Cleaning, Sterilization and Validation, Polycarbonate Base, Stainless Steel Needle, For Use With: For Cleaning, Sterilization and Validation, Quantity: 1 Each

  • Lower costs for cleaning, sterilization, and validation
  • Provide enhanced flexibility in batch scheduling and product development
  • Dimensional tolerances of +/-0.05% for assured repeatability
Specifications

Specifications

3.175 mm
For Cleaning, Sterilization and Validation
Stainless Steel
1 Each
Non-sterile
Dependent on pump used
Nozzle
101.6 mm
1/8 in.
Disposable filler nozzle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers