missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ Single-Use Pressure Transducer
Spectrum Labs™ Single-Use Pressure Transducer is designed to accurately measure feed, permeate, and retantate pressure during the tangential flow filtration process.
£155.00
Specifications
Accuracy | ±2% from 0 to 6 psi, ±3% from 6 to 30 psi |
---|---|
Material | Polysulfone |
Pressure Range | -10 to +75 psi |
Description
- Single-use
- Polysulfone construction
- Non-sterile
Specifications
±2% from 0 to 6 psi, ±3% from 6 to 30 psi | |
Polysulfone | |
-10 to +75 psi |