Learn More
STEMCELL Technologies SIS3
Activin/NODAL/TGF-β pathway inhibitor; Inhibits SMAD3
Brand:  STEMCELL Technologies 73962
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 17120302
  • £105.30 / 500µg
Estimated Shipment: 16-08-2024
to see stock.
Description
Description
  • SIS3 (specific inhibitor of SMAD3) is a cell-permeable selective inhibitor of transforming growth factor β1 (TGF-β1)-dependent SMAD3 phosphorylation (IC50 = 3 μM) and SMAD3-mediated cellular signaling (Jinnin et al.) SIS3 has no effect on SMAD2, p38 mitogen-activated protein kinase (MAPK), extracellular signal-regulated kinase (ERK), or phosphoinositide 3-kinases
  • Facilitates the production of anterior ectoderm from mouse induced pluripotent stem cells (Oshima et al.)
  • Inhibits myofibroblast differentiation of normal human dermal fibroblasts (Jinnin et al.)
Specifications
Specifications
1009104-85-1
500 μg
≥98%
Smad3 Inhibitor
C28H27N3O3