Learn More
STEMCELL Technologies SIS3
Activin/NODAL/TGF-β pathway inhibitor; Inhibits SMAD3
£105.30 - £191.70
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17120302
|
STEMCELL Technologies
73962 |
500 μg |
£105.30
500µg |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17130302
|
STEMCELL Technologies
73964 |
1 mg |
£191.70
1mg |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- SIS3 (specific inhibitor of SMAD3) is a cell-permeable selective inhibitor of transforming growth factor β1 (TGF-β1)-dependent SMAD3 phosphorylation (IC50 = 3 μM) and SMAD3-mediated cellular signaling (Jinnin et al.) SIS3 has no effect on SMAD2, p38 mitogen-activated protein kinase (MAPK), extracellular signal-regulated kinase (ERK), or phosphoinositide 3-kinases
- Facilitates the production of anterior ectoderm from mouse induced pluripotent stem cells (Oshima et al.)
- Inhibits myofibroblast differentiation of normal human dermal fibroblasts (Jinnin et al.)
Specifications
1009104-85-1 | |
C28H27N3O3 | |
≥98% |