missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SiteClick™ Antibody Labeling Kits

Generate brightly labeled antibodies for flow cytometry, fluorescence imaging, or western blot detection using SiteClick™ Qdot™ R-PE, and Biotin Antibody Labeling Kits.

Brand:  Invitrogen™ S10455

 View more versions of this product

Product Code. 15188735

  • £564.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Contains everything needed to azido modify and label 100 μg of antibody.
1 kit
SiteClick Labeling System
Wet Ice
Qdot 800
IgG
UV, Violet, Blue, Green, Yellow, Red
Labeling Kit
Store the entire kit at 2 to 6°C upon receipt
100 μg
Qdot™, SiteClick™
Near-IR
Qdot 525-800
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3071377 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog NumberS10455
Lot Number2971121 Certificate TypeCertificate of Analysis Date21/07/2024 Catalog NumberS10455
Lot Number2856040 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog NumberS10455
Lot Number2770807 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog NumberS10455
Lot Number2700988 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog NumberS10455
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ SiteClick™ Antibody Labeling Kits > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.