Learn More
Thermo Scientific™ Slide-A-Lyzer™ Accessory Float Buoys and Syringes
Slide-A-Lyzer Float Buoys and Syringes are available to support a variety of dialysis cassettes for volumes of 0.1 to 20 mL.
Brand: Thermo Scientific™ 66430
47.74 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : PN
Additional Details : Weight : 0.10000kg
Description
Slide-A-Lyzer Dialysis Cassette Syringes are designed for efficient loading of samples into Slide-A-Lyzer cassettes and removal of sample after dialysis. Cassette Float Buoy accessories are for use with Slide-A-Lyzer Dialysis Cassettes used to remove low molecular weight contaminants and perform buffer exchange, desalting, and concentration.
Minimize sample loss into and out of Slide-A-Lyzer Dialysis Cassettes using Slide-A-Lyzer Dialysis Cassette Syringes for 1-mL to 20-mL sample volumes. Slide-A-Lyzer Cassette Float Buoys are designed for floating 0.5- to 12-mL cassettes in dialysate.Features of Slide-A-Lyzer Cassette Accessories include:
• Syringes are appropriately sized for different sample volumes
• Buoys keep stir bar from tearing dialysis membrane
• Matched to Slide-A-Lyzer Cassette for a secure fit and minimization of sample loss
• Chemically inert
Specifications
Cassette Float Buoys (for 0.5–3 mL cassettes) | |
Store at room temperature. | |
Cassette, Accessory | |
10 Buoys |
Floating one 0.5 mL or 3 mL Cassette with each buoy | |
Dialysis | |
Slide-A-Lyzer™ | |
0.5 to 3 mL |