Learn More
Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 3.5K MWCO
Important Note: These G2 Dialysis Cassettes will be discontinued on June 1, 2023 and replaced by G3 Dialysis Cassettes, a new generation of cassettes. Consider switching to G3 Dialysis Cassettes. Remove salts and contaminants while maximizing sample recovery with these second-generation self-floating protein cassettes.
Brand: Thermo Scientific™ 87725
Description
Slide-A-Lyzer G2 Dialysis Cassettes provide maximum convenience, flexibility, and performance for sample dialysis. These 3.5K molecular weight cut off (3.5K MWCO) cassettes use a cellulose membrane to retain proteins and other macromolecules larger than 3,500 Daltons while allowing removal of salts and small contaminants. The cassette membrane is composed of low-binding regenerated cellulose. The membranes and cassettes are manufactured using clean-room conditions to ensure they are contaminant-free.
Slide-A-Lyzer G2 Dialysis Cassettes dialyze sample volumes up to 70 mL. These 3.5K MWCO cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.Features of Slide-A-Lyzer G2 Dialysis Cassettes (3.5K MWCO):
• Easy-loading—pipette-accessible for easy sample loading and retrieval
• Self-floating—integrated air chambers eliminate the need for float buoys
• Sturdy construction—ensures the highest possible sample integrity and protection
• Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
• Versatile—available for volumes of 0.5 to 70 mL
Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL
Order Info
Important Note: SKU #87725 will be discontinued on June 1, 2023 and replaced by A52968, a new generation of cassettes. Consider switching to A52968.
Specifications
Store at room temperature | |
Cellulose Membrane, Dialysis | |
3.5 kDa | |
6 Cassettes |
Buffer Exchange, Protein | |
Cassette | |
Slide-A-Lyzer™ | |
30 mL |