missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 3.5K MWCO

Important Note: These G2 Dialysis Cassettes will be discontinued on June 1, 2023 and replaced by G3 Dialysis Cassettes, a new generation of cassettes. Consider switching to G3 Dialysis Cassettes. Remove salts and contaminants while maximizing sample recovery with these second-generation self-floating protein cassettes.

£146.00 - £222.00

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 3.5 kDa
View More Specs

Products 5
Product Code Brand Sample Volume (Metric) Price Quantity & Availability  
Product Code Brand Sample Volume (Metric) Price Quantity & Availability  
10615655
View Documents
Thermo Scientific™
87722
0.5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10003594
View Documents
Thermo Scientific™
87723
3 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10515104
View Documents
Thermo Scientific™
87724
15 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10451375
View Documents
Thermo Scientific™
87725
30 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10378974
View Documents
Thermo Scientific™
87726
70 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Slide-A-Lyzer G2 Dialysis Cassettes provide maximum convenience, flexibility, and performance for sample dialysis. These 3.5K molecular weight cut off (3.5K MWCO) cassettes use a cellulose membrane to retain proteins and other macromolecules larger than 3,500 Daltons while allowing removal of salts and small contaminants. The cassette membrane is composed of low-binding regenerated cellulose. The membranes and cassettes are manufactured using clean-room conditions to ensure they are contaminant-free.

Slide-A-Lyzer G2 Dialysis Cassettes dialyze sample volumes up to 70 mL. These 3.5K MWCO cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.

Features of Slide-A-Lyzer G2 Dialysis Cassettes (3.5K MWCO):
Easy-loading—pipette-accessible for easy sample loading and retrieval
Self-floating—integrated air chambers eliminate the need for float buoys
Sturdy construction—ensures the highest possible sample integrity and protection
Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
Versatile—available for volumes of 0.5 to 70 mL

Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL

Specifications

Specifications

Store at room temperature
Cellulose Membrane, Dialysis
3.5 kDa
Buffer Exchange, Protein
Cassette
Slide-A-Lyzer™
Videos
SDS
Documents

Documents

Product Certifications