Learn More
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
15488881
|
Epredia™
A83900001 |
Slide Printer AS Basic |
£8555.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15408891
|
Epredia™
A83900002 |
Slide Printer AS Basic System |
£14160.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15418891
|
Epredia™
A83900004 |
Slide Printer AS On-Demand |
£11605.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15451031
|
Epredia™
A83900005 |
Slide Printer AS On-Demand Sys |
£26145.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SlideMate AS promotes single piece workflow though unique features designed specifically for on-demand printing.- Fully integrated color touch-screen and simple software are easy to use
- Built-in barcode scanner reads cassette information and imports the data to the slide label
- Connects easily to other computers or Laboratory Information Systems
- Prints directly onto the slide, eliminating the need for hand writing or labels
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Epredia™ SlideMate™ AS Slide Printer