missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ SlimLine™ Buckets with Casters
SlimLine™ narrow polypropylene buckets are for use with the SlimLine™ or Slim-T™ cleanroom cart system. Available in blue, red, green or yellow.
Brand: Micronova™ B-7REAC
Code : 03
Additional Details : Weight : 1.00000kg
Description
For use with SlimLine™ Cleanroom Cart System
- For use with SlimLine or SlimT Cleanroom Cart System
- 6 gallon capacity
- Autoclavable
- Available in blue, red, green or yellow
Specifications
SlimLine Bucket | |
Polypropylene Bucket, Nickel-plated Caster, Thermoplastic Rubber Wheel | |
6 gal. | |
Red | |
SlimLine or SlimT Cleanroom Cart System |
22.7 L | |
Yes | |
Store at Ambient Temperature. To Autoclave - do not stack - invert Bucket to allow drainage. | |
Cleaning Walls, Floors and Ceilings | |
Non-sterile |