missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ SlimLine™ Buckets with Casters
BIOPHARMA

SlimLine™ narrow polypropylene buckets are for use with the SlimLine™ or Slim-T™ cleanroom cart system. Available in blue, red, green or yellow.

Brand:  Micronova™ B-7DBAC

180.51 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 12871994

  • £242.00 / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with SlimLine™ Cleanroom Cart System

  • For use with SlimLine or SlimT Cleanroom Cart System
  • 6 gallon capacity
  • Autoclavable
  • Available in blue, red, green or yellow
Specifications

Specifications

SlimLine Bucket
Autoclavable
Dark Blue
SlimLine or SlimT Cleanroom Cart System
Polypropylene Bucket, Nickel-plated Caster, Thermoplastic Rubber Wheel
Store at Ambient Temperature. To Autoclave - do not stack - invert Bucket to allow drainage.
Cleaning Walls, Floors and Ceilings
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.