missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600
Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.
Brand: 3M™ Safety-Walk™ 7100146041
This item is not returnable.
View return policy
Description
- Anti-slip tape for light to heavy shoe-traffic areas
- High quality abrasive particles to ensure long-lasting friction in any application
- Highly durable surface withstands the rigors of equipment traffic
- High performance, pressure-sensitive adhesive for easy application
- Custom shapes and sizes are also available
- NFSI High Traction Certification

Specifications
18.3 m x 152 mm | |
Light to Heavy Traffic Areas | |
18.3 m | |
6 in. | |
Pressure Sensitive Adhesive | |
Floor Tape |
Mineral-coated | |
60 ft. | |
1 Ea. | |
152 mm | |
Transparent |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600 > 152 mm, Transparent
Spot an opportunity for improvement?Share a Content Correction