Learn More
Invitrogen™ SlowFade™ Gold Antifade Mountant
GREENER_CHOICE
Protects fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments.
Brand:  Invitrogen™ S36936
 View more versions of this product
Product Code. 11590676
  • £199.00 / 10mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides. It contains chemical components designed to protect fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments. SlowFade Gold Antifade Mountant is glycerol-based, with no mixing or curing required, which makes it ideal for immediate viewing of the sample. It comes ready-to-use—just apply a drop to the sample, add a coverslip, and image. It is available with or without DAPI nuclear stain.
Key attributes:
• Protects dyes from fading during imaging
• Ready-to-use liquid, ideal for immediate sample viewing
• Mounted samples are stable for weeks
• Maintains signal strength—little to no quenching
• Ideal for Alexa Fluor dyes
SlowFade Gold Antifade Mountant is not recommended for mounting samples containing fluorescent proteins, such as GFP. For superior antifade protection of fluorescent proteins and fluorescent dyes in cultured cells or thin tissue slices, SlowFade Diamond Antifade Mountant is recommended. For high resolution or to image thicker tissue or 3D cell culture with a focal depth of 0–500 μm, try SlowFade Glass Antifade Mountant.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Storage at room temperature is recommended but can also be stored frozen (-5° to -30°C). Protect from light.
SlowFade™
10 mL
Less hazardous, sustainable packaging
Liquid
Microscope
Mounting Media
Anti-fade Mountant
Room Temperature