Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin
Perform simple and easy streptavidin immunocapture for protein sample cleanup prior to protein digestion or analysis.
Brand: Thermo Scientific™ 60119-103
Description
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin. These kits were designed to remove the challenges often associated with the process of protein cleanup by providing a simple and easy capture workflow.
The kit contains beads with an attached streptavidin moiety enabling a strong streptavidin-biotin bond with the appropriate biotinylated molecule with specific affinity for the target biomolecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
Specifications
SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, SOLAμ Collection Plate, with Soluble Trypsin | |
C-terminal side of lysine and arginine amino acids | |
SMART Digest Resin, Bulk Magnetic | |
SMART Digest | |
Bulk Magnetic SMART Digest Resin, SMART Digest Soluble Trypsin, Collection Plate, SOLAμ SPE Plate |
Streptavidin | |
Trypsin | |
Immunocapture and Protein Digest Sample Preparation | |
Immunocapture + Protein Digestion (separate) |