Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin
Perform simple and easy streptavidin immunocapture for protein sample cleanup prior to protein digestion or analysis.
£678.38 - £1391.93
Specifications
Capture Streptavidin
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format SMART Digest Resin, Bulk Magnetic
Product Line SMART Digest
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
30130375
View Documents
Thermo Scientific™
60118-101
SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, without Soluble Trypsin
£678.38
1 set
30130372
View Documents
Thermo Scientific™
60119-103
SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, SOLAμ Collection Plate, with Soluble Trypsin
£1391.93
1 set
30130371
View Documents
Thermo Scientific™
60119-104
SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, with Soluble Trypsin
£1180.88
1 set
Description
Description
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin. These kits were designed to remove the challenges often associated with the process of protein cleanup by providing a simple and easy capture workflow.
The kit contains beads with an attached streptavidin moiety enabling a strong streptavidin-biotin bond with the appropriate biotinylated molecule with specific affinity for the target biomolecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
Specifications
Specifications
Streptavidin
Trypsin
SMART Digest
C-terminal side of lysine and arginine amino acids
SMART Digest Resin, Bulk Magnetic
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin