Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin
Perform simple and easy streptavidin immunocapture for protein sample cleanup prior to protein digestion or analysis.
Brand: Thermo Scientific™ 60118-101
Description
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin. These kits were designed to remove the challenges often associated with the process of protein cleanup by providing a simple and easy capture workflow.
The kit contains beads with an attached streptavidin moiety enabling a strong streptavidin-biotin bond with the appropriate biotinylated molecule with specific affinity for the target biomolecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
Specifications
SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, without Soluble Trypsin | |
C-terminal side of lysine and arginine amino acids | |
SMART Digest Resin, Bulk Magnetic | |
SMART Digest | |
Bulk Magnetic SMART Digest Resin |
Streptavidin | |
Trypsin | |
Immunocapture Sample Preparation | |
Immunocapture |