Learn More
Thermo Scientific™ SMART SPME Arrows for TriPlus™ RSH SMART Autosamplers
Track part and lot numbers, usage range, history, and more for Thermo Scientific™ SMART SPME Arrows on the TriPlus™ RSH SMART Autosampler.
£456.94 - £2218.28
Product Code | Brand | Description | Color | Diameter (Metric) Outer | Phase | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Color | Diameter (Metric) Outer | Phase | Price | Quantity & Availability | ||
17767444
|
Thermo Scientific™
36SA10A1-SM |
SMART SPME Arrow 1.1 mm: Acrylate (Polyacrylate), 100 μm, Grey, 1/Pk. | Gray | 1.1 mm | Acrylate (Polyacrylate), 100 μm |
£456.94
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||
17777444
|
Thermo Scientific™
36SA10A3-SM |
SMART SPME Arrow 1.1 mm: Acrylate (Polyacrylate), 100 μm, Grey, 3/Pk. | Gray | 1.1 mm | Acrylate (Polyacrylate), 100 μm |
£1273.52
Pack of 3 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
17787444
|
Thermo Scientific™
36SA10M5-SM |
SMART SPME Arrows 1.1 mm: Collection of Five Different SMART SPME Arrows | Gray/Light Blue/Violet/Red/Dark Gray | 1.1 mm | Acrylate Thickness 100 μm; Carbon WR/PDMS Thickness 120 μm; DVB/PDMS Thickness 120 μm; PDMS Thickness 100 μm; DVB/Carbon WR/PDMS Thickness 120 μm |
£2218.28
Pack of 5 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
17797444
|
Thermo Scientific™
36SA10P1-SM |
SMART SPME Arrow 1.1 mm: PDMS (Polydimethylsiloxane), 100 μm, Red, 1/Pk. | Red | 1.1 mm | PDMS (Polydimethylsiloxane), 100 μm |
£456.94
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
17707454
|
Thermo Scientific™
36SA10P3-SM |
SMART SPME Arrow 1.1 mm: PDMS (Polydimethylsiloxane), 100 μm, Red, 3/Pk. | Red | 1.1 mm | PDMS (Polydimethylsiloxane), 100 μm |
£1273.52
Pack of 3 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
17717454
|
Thermo Scientific™
36SA11T1-SM |
SMART SPME Arrow 1.1 mm: DVB/C-WR/PDMS (Divinylbenzene/Carbon WR/Polydimethylsiloxane), 120 μm, Dark Grey, 1/Pk. | Dark Gray | 1.1 mm | DVB/C-WR/PDMS (Divinylbenzene/Carbon WR/Polydimethylsiloxane), 120 μm |
£456.94
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
17727454
|
Thermo Scientific™
36SA11T3-SM |
SMART SPME Arrow 1.1 mm: DVB/C-WR/PDMS (Divinylbenzene/Carbon WR/Polydimethylsiloxane), 120 μm, Dark Grey, 3/Pk. | Dark Gray | 1.1 mm | DVB/C-WR/PDMS (Divinylbenzene/Carbon WR/Polydimethylsiloxane), 120 μm |
£1273.52
Pack of 3 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
17737454
|
Thermo Scientific™
36SA12B1-SM |
SMART SPME Arrow 1.1 mm: Carbon WR/PDMS (Carbon Wide Range), 120 μm, Light Blue, 1/Pk. | Light Blue | 1.1 mm | Carbon WR/PDMS (Carbon Wide Range/ Polydimethylsiloxane), 120 μm |
£456.94
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
17747454
|
Thermo Scientific™
36SA12B3-SM |
SMART SPME Arrow 1.1 mm: Carbon WR/PDMS (Carbon Wide Range), 120 μm, Light Blue, 3/Pk. | Light Blue | 1.1 mm | Carbon WR/PDMS (Carbon Wide Range/ Polydimethylsiloxane), 120 μm |
£1273.52
Pack of 3 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
17757454
|
Thermo Scientific™
36SA12E1-SM |
SMART SPME Arrow 1.1 mm: DVB/PDMS (Divinylbenzene), 120 μm, Violet, 1/Pk. | Violet | 1.1 mm | DVB/PDMS (Divinylbenzene), 120 μm |
£456.94
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
17767454
|
Thermo Scientific™
36SA12E3-SM |
SMART SPME Arrow 1.1 mm: DVB/PDMS (Divinylbenzene), 120 μm, Violet, 3/Pk. | Violet | 1.1 mm | DVB/PDMS (Divinylbenzene), 120 μm |
£1273.52
Pack of 3 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
17777454
|
Thermo Scientific™
36SA25P1-SM |
SMART SPME Arrow 1.5 mm: PDMS (Polydimethylsiloxane), 250 μm, Black, 1/Pk. | Black | 1.5 mm | PDMS (Polydimethylsiloxane), 250 μm |
£548.34
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
17787454
|
Thermo Scientific™
36SA25P3-SM |
SMART SPME Arrow 1.5 mm: PDMS (Polydimethylsiloxane), 250 μm, Black, 3/Pk. | Black | 1.5 mm | PDMS (Polydimethylsiloxane), 250 μm |
£1527.52
Pack of 3 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
Description
Track information such as part number, lot number, usage range, and history for Thermo Scientific™ SMART SPME Arrows installed on TriPlus™ RSH SMART Autosamplers. Each arrow is equipped with an ID chip, which stores this usage-based information for fast, solvent-free, and automated sample preparation. When installed on the TriPlus RSH SMART Autosampler, the information for SMART SPME Arrows can be directly accessed and reported via Thermo Scientific™ Chromeleon™ CDS software.
Automate solvent-free solid phase micro extraction (SPME) sample preparation by performing sample extraction and enrichment in a single step with our SMART SPME Arrows.
SMART SPME Arrows also offer the additional benefit of tracking usage information. SMART SPME Arrows are compatible with all TriPlus™ RSH instruments as well as PAL platforms and AOC-6000 series instruments.
SMART Technology Benefits
- Gain higher confidence in analytical results
- Ensure error free usage of SPME Arrows through automatic identification
- Support GLP accreditation with usage history traceability
- Optimize consumption by monitoring of usage conditions and lifetime
- Facilitate SPME Arrow management thanks to color-coded health notification dashboard integrated in Chromeleon™ CDS
Specifications
Phase Length: 20 mm | |
Arrow |