Learn More
Eppendorf™ SmartBlock DWP500
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
Deepwell plates, 96/500 μL
Description
- For Eppendorf Deepwell Plate 96/500 μL
- Optimized block design supports optimal temperature transfer within your sample to receive excellent results
- Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
- Every SmartBlock is individually inspected in production for temperature accuracy and is delivered includes serial-number-specific certificate for documentation purpose
- Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept)
- Automatic block recognition for safe handling
- Eppendorf QuickRelease™ technology enables fast and simple block exchange - no need for tools
- Limited to ThermoMixer C and ThermoStat C
Specifications
Specifications
For Use With (Equipment) | Deepwell plates, 96/500 μL |
Type | Block |
Includes | Lid |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
9Minutes
44Seconds
2
Product Content Correction
Product Title
Eppendorf™ SmartBlock DWP500 >
Spot an opportunity for improvement?Share a Content Correction