Learn More
ICU Medical Jelco™ Saf-T Holder™ Blood Culture Holders and Transfer Devices
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
POA
Specifications
For Use With (Application) | Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw |
---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
15795068
![]() |
ICU Medical
96005 |
Saf-T Holder™ Blood Culture Device, with female luer |
POA
Pack of 50 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Eliminate potential exposures and holder reuse
- Provide safe needleless transfer after a syringe draw
Specifications
Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.