missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Smooth Folded Filters, Grade 3-HW
Use for routine qualitative analyses in the lab. Sartorius™ Smooth Folded Filters, Grade 3-HW are wet-strengthened for use in pressure and vacuum filtration. Ultra-pure filter paper is available as a folded filter cone in a variety of diameters.
£36.30 - £493.00
Specifications
Format | Folded |
---|---|
Material | Cotton Linters, Refined Pulp |
Particle Retention | 8 to 12 μm |
Filtration Speed | 20μm/sec. |
For Use With (Application) | For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications |
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | |||||
10672592
|
Sartorius™
FT-4-303-070 |
70 mm |
£287.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10468742
|
Sartorius™
FT-4-303-110 |
110 mm |
£50.60
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10642042
|
Sartorius™
FT-4-303-125 |
125 mm |
£36.30
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10233381
|
Sartorius™
FT-4-303-150 |
150 mm |
£36.30
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10480663
|
Sartorius™
FT-4-303-185 |
185 mm |
£36.30
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10355171
|
Sartorius™
FT-4-303-090 |
90 mm |
£112.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10173941
|
Sartorius™
FT-4-303-210 |
210 mm |
£48.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10662422
|
Sartorius™
FT-4-303-240 |
240 mm |
£61.80
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10479891
|
Sartorius™
FT-4-303-290 |
290 mm |
£65.80
Pack of 100 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10375411
|
Sartorius™
FT-4-303-320 |
320 mm |
£89.50
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10032291
|
Sartorius™
FT-4-303-385 |
385 mm |
£226.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10134041
|
Sartorius™
FT-4-303-450 |
450 mm |
£493.00
Pack of 100 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
10286842
|
Sartorius™
FT-4-303-500 |
500 mm |
£287.00
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
Description
These smooth filter paper folded cones are used for routine qualitative analyses, such as clarification and determination of substances. Filters are wet-strengthened, so they are suitable for pressure or vacuum filtration.- Made of refined pulp and cotton linters with a α-cellulose content greater than 95%
- Pure, technical filter paper with an ash content of 0.1 to 0.15%
- Grade 3-HW filters feature the following characteristics:
- Medium to wide pore for a moderately-fast filtering rate
- Wet burst resistance of 15kPa
- Basis weight of 65g/m2
- Non-sterile and bulk packed
Specifications
Folded | |
8 to 12 μm | |
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications | |
100 Pack | |
Qualitative, Technical, Smooth |
Cotton Linters, Refined Pulp | |
20μm/sec. | |
3HW | |
0.14 mm |