missing translation for 'onlineSavingsMsg'
Learn More

Motic™ SMZ-168 Microscope Head

Replacement or modular head suitable for the SMZ-168 Stereo Microscope. Motic™ SMZ-168 Microscope Head is a core element of a Motic™ microscope with one of the industry's largest zoom ratios.

Brand:  Motic™ PX55000001

 View more versions of this product

Product Code. 11800113

  • £793.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The SMZ-168 Stereo Zoom Microscope represents the top-of-the-line model in the SMZ Series.
  • Designed for a wide range of biological and material science applications.
  • High optical quality and versatile accessories satisfy the most demanding user applications in both routine and research fields.
  • Durable and suitable for a variety of applications.
  • Greenough optical system comes with a large zoom range of 6.7:1, parfocal at all magnifications.
  • Five-level position click-stop magnification mechanism, implemented into the continuous zoom, enables precise reproduction of magnifications, thus simplifying measurements.
  • Impressive 3-dimensional upright image, combined with an industry leading working distance of 113mm in the standard configuration, allows easy sample manipulation.
  • Converts easily into a complete documentation station by combining it with a Moticam camera.
Specifications

Specifications

0.75X to 5X
6.7:1 Zoom ratio
SMZ-168 TH
Widefield WF10X/23 mm, 100:0 (Eyetubes), 0:100 (Left Eyetube/Trinocular Tube) Optical Split
SMZ-168 Microscopes
Microscope Head
Trinocular, 35° Inclined
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.