Learn More
Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes

Improve sample recovery by significantly reducing surface binding with these tubes that do not have additives or coatings.
Brand: Thermo Scientific™ 3446
25.30 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Improve sample recovery by significantly reducing surface binding with Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes. Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. With no additives or coatings, these tubes — available in 0.6mL, 1.5mL and 2.0mL sizes — offer maximum sample recovery by significantly reducing surface binding.
- The duckbill cap reduces the pressure required to open the tube with a wider contact surface, yet ensures the cap will remain closed during centrifugation
- Low retention properties are not achieved by a coating or plastic-embedding processes, ensuring that samples will not be contaminated from these additives
- Tubes will remain closed during centrifugation yet open easily with one hand as needed
- Tubes are available in amber to protect your light-sensitive samples
- Graduation marks on the microcentrifuge tube allow for effective sample reference
Color/Packaging Options
- A variety of packaging options are available to help with contamination control in the lab
- For stringent sterility requirements, individually wrapped sterile tubes are available
- Color options, in addition to natural and amber, are red, blue, green, yellow and violet and a mixed color packaging
Highest Quality
- Manufactured from virgin polypropylene, these microcentrifuge tubes ensure sample integrity during boiling, freezing and centrifugation
- Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
- Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Recommended for:
Daily laboratory usage including centrifugation and short-term sample storage

Specifications
Flat | |
0.6 mL | |
Polypropylene | |
Non-sterile | |
30,000 x g |
Certified free of RNase, DNase, DNA and pyrogens | |
Natural | |
100 tubes/ bag, 10 bags/ unit, 10 units/ case | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.