missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SNaPshot™ Multiplex Kit

The SNaPshot™ Multiplex Kit is part of the SNaPshot™ Multiplex System. The SNaPshot™ Multiplex System primer extension-based method enables multiplexing up to 10 single nucleotide polymorphisms (SNPs).

Brand:  Applied Biosystems™ 4323151

Additional Details : Weight : 0.08160kg

 View more versions of this product

Product Code. 15999172

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: SNaPshot™ multiplex ready reaction mix (for 100 rxns), SNaPshot™ multiplex control primer mix (30μL), SNaPshot™ multiplex control template (60μL), protocol

Description

Description

  • Perform multiplexed, single-base extension of up to 10 SNPs
  • Interrogate multiple SNPs regardless of chromosome position
  • Separate SNP loci that differ by a single base pair
  • Perform robust interrogation of multiple SNPs with only a small amount of amplified template
  • Decrease expenses with this moderately priced system
  • Reduce the complexity of your experiments with this well-tested, easy-to-understand single-base extension technique

Order Info

Each

Specifications

Specifications

Contains:
• SNaPshot™ Multiplex Ready Reaction Mix (for 100 reactions)
• SNaPshot™ Multiplex Control Primer Mix (30 μl)
• SNaPshot™ Multiplex Control Template (60 μl)
• Protocol

Store at -15°C to -25°C in a constant-temperature freezer.
Kit
SNP Genotyping (Fragment Analysis-Based), Primer Extension
SNaPshot™
9 hr.
SNPs (Known)
100 Reactions, with protocol
Primer-probe
9 hr.
3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer, 3730/3730xl Genetic Analyzers
Multiplex Kit
All Species
Genotyping
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.