missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SNaPshot™ Multiplex Kit

The SNaPshot™ Multiplex Kit is part of the SNaPshot™ Multiplex System. The SNaPshot™ Multiplex System primer extension-based method enables multiplexing up to 10 single nucleotide polymorphisms (SNPs).

Brand:  Applied Biosystems™ 4323161

Additional Details : Weight : 0.04310kg

 View more versions of this product

Product Code. 16355282

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: SNaPshot™ Multiplex Ready Reaction Mix (for 1000 reactions), SNaPshot™ Multiplex Control Primer Mix (30μL), SNaPshot™ Multiplex Control Template (60μL)

Description

Description

  • Interrogate up to 10 SNPs from different amplicons in a single base extension reaction
  • Separate SNP loci that differ by a single base pair
  • Increased sensitivity compared to standard sequencing allows detection of minor variants (e.g. somatic mutations) in a test sample
  • Reduce the complexity of your experiments with this easy to use and cost-efficient genotyping system
  • A large body of scientific literature demonstrates the utility for genotyping, mutation detection and mapping

Order Info

Each

Specifications

Specifications

Contains:
• SNaPshot™ Multiplex Ready Reaction Mix (for 1000 reactions)
• SNaPshot™ Multiplex Control Primer Mix (30 μl)
• SNaPshot™ Multiplex Control Template (60 μl)

Store at -15°C to -25°C in a constant-temperature freezer.
Primer-probe
AmpliTaq DNA Polymerase
3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer, 3730/3730xl Genetic Analyzers
SNaPshot™
Dry Ice
SNPs (Known)
No
DNA
SNaPshot™ Multiplex Kit, 1000 rxns, Capillary Electrophoresis Detection Method, DNA Sample Type, AmpliTaq™ DNA Polymerase, SNPs (Known) Genotyping Target, Multiplexing High Throughput Compatibility, Unlabelled Oligos Primer-Probe Compatibility
Tube
SNP Genotyping (Fragment Analysis-Based), Primer Extension
Multiplexing
Multiplex Kit
GeneMapper™ Software
Genotyping
1000 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.