missing translation for 'onlineSavingsMsg'
Learn More

Jaytec Glass™ Soda Glass Melting Point Tubes

Heat samples in Soda Glass Melting Point Tubes to determine their melting point. These tubes are made of industry-standard soda lime glass.

Brand:  Jaytec Glass™ CAP-MPC-75-1.8

42.54 GBP valid until 2025-01-17
Use promo code "21645" to get your promotional price.


 View more versions of this product

Product Code. 15279628

  • £78.40 / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Designed to be compatible with all major melting point apparatus
  • Available in 75 mm or 100 mm lengths
Specifications

Specifications

Melting Point Instrument
75 mm
One end sealed
1.8 to 1.9 mm
Soda Lime Glass
1000 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.