missing translation for 'onlineSavingsMsg'
Learn More

Soda Glass Test tubes

Use these test tubes for general mixing and simple laboratory work. Soda Glass Test tubes will withstand moderate warming (ex. in a water bath).

Brand:  Samco G001/72

 View more versions of this product

Product Code. 12319329

  • £95.95 / Pack of 250
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Light walled test tube with rim
  • Should not be placed directly into naked flames
  • Available in packs of 250
Specifications

Specifications

ISO 4142:2002
General mixing, Simple laboratory work
Not Autoclavable
50 mm
Rim
10 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.