missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Soda Lime, Extra Pure, SLR, 2.5-5mm, Not Hygroscopic, Granules, Fisher Chemical™
£37.95 - £224.00
Chemical Identifiers
CAS | 8006-28-8 |
---|---|
Molecular Formula | CaHNaO2 |
Molecular Weight (g/mol) | 96.074 |
MDL Number | 134094 |
InChI Key | HUAUNKAZQWMVFY-UHFFFAOYSA-M |
Synonym | soda lime, soda-lime, sodasorb, soda lime nf, sodium hydroxide na oh , mixt. with lime, soda-lime, granular, soda lime, indicating, soda lime, indicator grade, soda lime with sodium hydroxide, soda lime, indicating 8-12 mesh |
PubChem CID | 66545795 |
IUPAC Name | sodium;oxocalcium;hydroxide |
SMILES | [OH-].O=[Ca].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12636767
|
Fisher Chemical
S/1560/50 |
250 g | Plastic powder jar | N/A | |||||
10549200
|
Fisher Chemical
S/1560/53 |
500 g | Plastic powder jar | N/A | |||||
12646767
|
Fisher Chemical
S/1560/60 |
1 kg | Plastic powder jar | N/A | |||||
10470384
|
Fisher Chemical
S/1560/65 |
5 kg | Plastic keg | N/A | |||||
Description
Soda Lime, Extra Pure, SLR, 2.5-5mm, Not Hygroscopic, Granules
Chemical Identifiers
8006-28-8 | |
96.074 | |
HUAUNKAZQWMVFY-UHFFFAOYSA-M | |
66545795 | |
[OH-].O=[Ca].[Na+] |
CaHNaO2 | |
134094 | |
soda lime, soda-lime, sodasorb, soda lime nf, sodium hydroxide na oh , mixt. with lime, soda-lime, granular, soda lime, indicating, soda lime, indicator grade, soda lime with sodium hydroxide, soda lime, indicating 8-12 mesh | |
sodium;oxocalcium;hydroxide |
Specifications
8006-28-8 | |
Solid | |
134094 | |
HUAUNKAZQWMVFY-UHFFFAOYSA-M | |
sodium;oxocalcium;hydroxide | |
66545795 |
White | |
CaHNaO2 | |
soda lime, soda-lime, sodasorb, soda lime nf, sodium hydroxide na oh , mixt. with lime, soda-lime, granular, soda lime, indicating, soda lime, indicator grade, soda lime with sodium hydroxide, soda lime, indicating 8-12 mesh | |
[OH-].O=[Ca].[Na+] | |
96.074 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Soda lime
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.