missing translation for 'onlineSavingsMsg'
Learn More

Soda Lime Glass Centrifuge Tubes with Rim

Use these tubes for a variety of applications. Soda Lime Glass Centrifuge Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.

Brand:  Fisherbrand G132/01

 View more versions of this product

Product Code. 15925720

  • £30.40 / Pack of 10
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Soda glass
  • Capacity 15mL
  • Max. RCF x g: 1300
Specifications

Specifications

15 mL
100
Conical
Yes
1300 x g
Soda Lime Glass
Conical
Yes
Centrifuge Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.