missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Soda Lime Glass Culture Tubes

Manufactured from soda lime glass. BRAND™ Soda Lime Glass Culture Tubes are rimless and autoclavable at 121°C.

Brand:  Brand™ 114115

 View more versions of this product

Product Code. 12209309

This item is not returnable. View return policy

Description

Description

  • Manufactured from soda lime glass.
  • Rimless
  • Autoclavable at 121°C.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mm
78/Pk.
Culture Tube
Soda Lime Glass
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers