missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Soda Lime Glass Culture Tubes

Manufactured from soda lime glass. BRAND™ Soda Lime Glass Culture Tubes are rimless and autoclavable at 121°C.

Brand:  Brand™ 114110

 View more versions of this product

Product Code. 12626215

  • £21.60 / Pack of 144
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from soda lime glass.
  • Rimless
  • Autoclavable at 121°C.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mm
144/Pk.
Culture Tube
Soda Lime Glass
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.