missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube

Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.

Brand:  Fisherbrand™ FB63107

 View more versions of this product

Product Code. 11962178

This item is not returnable. View return policy

Description

Description

  • Features: rimmed light walls
  • Withstands: moderate warming
  • Complies to: ISO 4142
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 4142
Soda Lime Glass
Yes
150 mm
50 Pack
24 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube > Dimensions: 24 dia. x 150mmH

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.