Learn More
BRAND™ Soda Lime Glass Petri Dishes
Perfect for microbial or cell culture applications, these Soda Lime Glass Petri Dishes feature excellent glass quality and workmanship.
Brand:  BRAND™ 455732
30.13 GBP valid until 2024-12-31
Use promo code "21955" to get your promotional price.
 View more versions of this product
Product Code. 10315571
  • £41.00 / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Dish and lid are flat inside and outside
  • Free from bubbles and streaks
  • Cut edges are fire-polished
  • Suitable for short-term chemical exposure
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Soda Lime Glass
Fire-polished cut edges
80 mm
Petri Dish
15 mm