missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate 7.5% solution
GREENER_CHOICE

Sodium Bicarbonate 7.5% solution

Brand:  Gibco 25080102

 View more versions of this product

Product Code. 11550626

  • £143.00 / Pack of 20

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Sodium Bicarbonate is buffer commonly used for maintaining the pH of cell culture medium in the presence of 4–10% carbon dioxide. In addition to buffering, sodium bicarbonate provides some nutritional benefit, while rarely displaying any cell toxicity. Formulated as 75 g per liter of water, Gibco™ Sodium Bicarbonate, 7.5% Solution is perfect for supplementing dry powder medium during reconstitution. The final Sodium Bicarbonate concentration used in the medium depends on the media formulation and the carbon dioxide concentration used in the incubator.

Product Use
For Research Use Only. Not for use in diagnostic procedures.

Dual-site cGMP Manufacturing and Quality System
Gibco™ Sodium Bicarbonate Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Sodium Bicarbonate Solution product made in our Grand Island facility (25080-094). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

15 to 30°C
7.5 %
Sodium Bicarbonate
20 x 100 mL
Sodium Bicarbonate 7.5% Solution, 20 x 100mL, 7.5% Concentration, Liquid, Buffer, Used in Mammalian Cell Culture, Perfect for Supplementing Dry Powder Medium during Reconstitution, For Research Use Only, Not for use in Diagnostic Procedures
Liquid
Mammalian Cell Culture
For research use only
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.