missing translation for 'onlineSavingsMsg'
Learn More
Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals Product Code.: 10583381

Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals

Product Code. 10583381
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10583381

Brand: Thermo Scientific Acros 217125000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neutShow MoreShow Less
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 99.50%
Linear Formula NaHCO3
Fieser 03,260; 05,595; 11,476; 17,180
Merck Index 15, 8719
Solubility Information Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium bicarbonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467945 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number217125000,217120100,217120010,217120025
Lot NumberA0467761 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number217125000,217120100,217120010,217120025
Lot NumberA0463573 Certificate TypeCertificate of Analysis Date21/06/2024 Catalog Number217125000,217120100,217120010,217120025
Lot NumberA0463611 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number217125000,217120100,217120010,217120025
Lot NumberA0458755 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog Number217125000,217120100,217120010,217120025
Product Identifier
  • Sodium bicarbonate
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    1

    Minutes

    48

    Seconds

    3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.