missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals
£21.00 - £188.94
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neutShow More |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
SMILES | [Na+].OC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10583381
|
Thermo Scientific Acros
217125000 |
500 g | Plastic bottle |
£21.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10489910
|
Thermo Scientific Acros
217120010 |
1 kg | Plastic bottle |
£32.04
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10082840
|
Thermo Scientific Acros
217120025 |
2.5 kg | Plastic bottle |
£60.17
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10144713
|
Thermo Scientific Acros
217120100 |
10 kg | Plastic drum |
£188.94
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
516892 | |
[Na+].OC([O-])=O |
CHNaO3 | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neutShow More | |
CHEBI:32139 |
Specifications
270.0°C | |
White | |
99.50% | |
NaHCO3 | |
03,260; 05,595; 11,476; 17,180 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
84.01 | |
Analytical |
144-55-8 | |
Fine Crystalline Powder | |
CHNaO3 | |
MFCD00003528 | |
15, 8719 | |
Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 | |
99.5% | |
Sodium bicarbonate |
Safety and Handling
missing translation for 'einecsNumber' : 205-633-8
Product Identifier
- Sodium bicarbonate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals