missing translation for 'onlineSavingsMsg'
Learn More
Sodium bicarbonate, 99+%, for HPLC, Thermo Scientific Chemicals Product Code.: 11904311

Sodium bicarbonate, 99+%, for HPLC, Thermo Scientific Chemicals

Product Code. 11904311
250 g, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11904311

Brand: Thermo Scientific Acros 446232500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neutShow MoreShow Less
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270.0°C
Quantity 250 g
Assay Percent Range 99+%
Linear Formula NaHCO3
Merck Index 15,8719
Solubility Information Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol
IUPAC Name sodium hydrogen carbonate
Formula Weight 84.01
Percent Purity ≥99%
Grade HPLC
Packaging Glass bottle
Chemical Name or Material Sodium bicarbonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469047 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number446232500,446230010
Lot NumberA0442580 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number446232500,446230010
Lot NumberA0456584 Certificate TypeCertificate of Analysis Date28/09/2023 Catalog Number446232500,446230010
Lot NumberA0431964 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number446232500,446230010
Lot NumberA0393937 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number446232500,446230010
Product Identifier
  • Sodium bicarbonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium bicarbonate, 99+%, for HPLC, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.