missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, Honeywell Fluka™

ACS Reagent,≥99.7%

£24.36 - £822.57

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O
View More Specs

Products 9
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15638100
View Documents
Honeywell-Fluka
S6014-25G
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15658100
View Documents
Honeywell-Fluka
S6014-500G
500 g
£52.19
500g
Please to purchase this item. Need a web account? Register with us today!
15618100
View Documents
Honeywell-Fluka
S6014-1KG
1 kg
£40.47
1kg
Please to purchase this item. Need a web account? Register with us today!
15628100
View Documents
Honeywell-Fluka
S6014-2.5KG
2.5 kg
£89.97
2.50kg
Please to purchase this item. Need a web account? Register with us today!
15688100
View Documents
Honeywell-Fluka
S6014-6X500G
6 x 500 g
£172.97
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15678100
View Documents
Honeywell-Fluka
S6014-5KG
5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15648100
View Documents
Honeywell-Fluka
S6014-4X2.5KG
4 x 2.5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15608100
View Documents
Honeywell-Fluka
S6014-12KG
12 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15668100
View Documents
Honeywell-Fluka
S6014-50KG
50 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

144-55-8
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
[Na+].OC([O-])=O
CHNaO3
MFCD00003528
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
CHEBI:32139

Specifications

144-55-8
NaHCO3
NONH for all modes of transport
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
[Na+].OC([O-])=O
84.01
CHEBI:32139
99.7 to 100.3%
CHNaO3
MFCD00003528
4153970
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium;hydrogen carbonate
516892
84.01g/mol
Sodium bicarbonate
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : VZ0950000

SDS
Documents

Documents

Product Certifications