missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
£16.78 - £214.07
Chemical Identifiers
CAS | 7647-15-6 |
---|---|
Molecular Formula | BrNa |
Molecular Weight (g/mol) | 102.89 |
MDL Number | MFCD00003475 |
InChI Key | JHJLBTNAGRQEKS-UHFFFAOYSA-M |
Synonym | sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a |
PubChem CID | 253881 |
ChEBI | CHEBI:63004 |
IUPAC Name | sodium bromide |
SMILES | [Na+].[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10216740
|
Thermo Scientific Acros
205131000 |
100 g | Plastic bottle |
£16.78
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10368100
|
Thermo Scientific Acros
205130010 |
1 kg | Plastic bottle |
£59.90
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10063573
|
Thermo Scientific Acros
205130050 |
5 kg | Plastic bottle |
£214.07
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7647-15-6 | |
102.89 | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
253881 | |
sodium bromide |
BrNa | |
MFCD00003475 | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
CHEBI:63004 | |
[Na+].[Br-] |
Specifications
1390.0°C | |
7647-15-6 | |
5 to 7 (5% soln. at 20°C) | |
99% min. (on dry substance) (Argentometry) | |
NaBr | |
12,445; 13,46 | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
sodium bromide | |
253881 | |
102.89 | |
Extra Pure | |
Sodium bromide, Anhydrous |
755.0°C | |
White | |
Crystalline Powder | |
BrNa | |
MFCD00003475 | |
15, 8729 | |
Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol | |
[Na+].[Br-] | |
102.89 | |
CHEBI:63004 | |
≥99% | |
1% max. (120°C) |
Safety and Handling
missing translation for 'einecsNumber' : 231-599-9
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title