missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Monohydrate, Crystal, N.F. - F.C.C., J.T. Baker™
£145.00 - £635.00
Chemical Identifiers
CAS | 5968-11-6 |
---|---|
Molecular Formula | CH2Na2O4 |
Molecular Weight (g/mol) | 124.00 |
InChI Key | MQRJBSHKWOFOGF-UHFFFAOYSA-L |
Synonym | sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajopShow More |
PubChem CID | 2735133 |
IUPAC Name | disodium;carbonate;hydrate |
SMILES | C(=O)([O-])[O-].O.[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15568144
|
Avantor J.T.Baker
3600-01 |
500 g | Poly Bottle |
£145.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15578144
|
Avantor J.T.Baker
3600-05 |
2.5 kg | Poly Bottle |
£635.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
5968-11-6 | |
124.00 | |
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajopShow More | |
disodium;carbonate;hydrate |
CH2Na2O4 | |
MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
2735133 | |
C(=O)([O-])[O-].O.[Na+].[Na+] |
Specifications
5968-11-6 | |
CH2Na2O4 | |
MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
disodium;carbonate;hydrate | |
2735133 | |
FCC/NF | |
Sodium Carbonate, Monohydrate |
Crystals | |
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
C(=O)([O-])[O-].O.[Na+].[Na+] | |
124.00 | |
124 | |
1L = 2.25kg |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium carbonate monohydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sodium carbonate monohydrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Carbonate, Monohydrate, Crystal, N.F. - F.C.C., J.T. Baker™