Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium chloride, 99.5%, for analysis, Thermo Scientific Chemicals
Sodium chloride, 99.5%, for analysis, Thermo Scientific Chemicals
Product Code. 10216410Product Code. 10216410
Brand: Thermo Scientific Acros 207790050

Use promo code "24155" to get your promotional price.
Explore more special offers
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Chemical Identifiers
CAS | 7647-14-5 |
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
IUPAC Name | sodium;chloride |
SMILES | [Na+].[Cl-] |
Specifications
Boiling Point | 1461.0°C |
Melting Point | 801.0°C |
Color | White |
pH | 5.0 to 8.0 (5% soln. at 20°C) |
Physical Form | Fine Crystalline Powder |
Quantity | 5 kg |
Assay Percent Range | 99.4% min. (Argentometry) |
Linear Formula | NaCl |
Fieser | 13,280 |
Merck Index | 15, 8734 |
Solubility Information | Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid |
Formula Weight | 58.44 |
Percent Purity | 99.5% |
Grade | Analytical |
Packaging | Plastic Bottle |
Chemical Name or Material | Sodium chloride |
Show More Show Less |
Product Suggestions
You may be interested in these alternatives
-
Product Code. 10194763
-
Product Code. 10428420
-
Product Code. 10514283
-
Product Code. 15631310
-
Product Code. 12685077
-
Product Code. 10478283
-
Product Code. 15673410
-
Product Code. 15646770
-
Product Code. 10092740
-
Product Code. 10378573
Customers who viewed this item also viewed. This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
-
Catalog No. 10112640
£50.40 / 1kg -
Catalog No. 10055850
£21.91 / 500g -
Catalog No. 10428420
£72.05 / 1kg -
Catalog No. 10316943
£56.15 / 1kg -
Catalog No. 15526005
£32.15 / 500g -
Catalog No. 12675077
£38.08 / 1kg -
Catalog No. 11984051
£97.35 / 1kg -
Catalog No. 10675112
£48.80 / 2.50 L -
Catalog No. 10316380
£75.90 / 2.50 L -
Catalog No. 10159872
£47.56 / 1kg -
Catalog No. 10660131
£315.00 / 2.50 L -
Catalog No. 11462858
£23.70 / 500g -
Catalog No. 10376743
£169.00 / 1kg
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Lot Number2497961 | Certificate TypeCertificate of Analysis | Date31/01/2025 | Catalog Number1093/E,207790050,207790250,207790010 |
Lot Number2404508 | Certificate TypeCertificate of Analysis | Date31/01/2025 | Catalog Number1093/E,207790050,207790250,207790010 |
Lot NumberA0468368 | Certificate TypeCertificate of Analysis | Date31/01/2025 | Catalog Number1093/E,207790050,207790250,207790010 |
Lot Number2407894 | Certificate TypeCertificate of Analysis | Date30/01/2025 | Catalog Number1093/E,207790050,207790250,207790010 |
Lot Number2192379 | Certificate TypeCertificate of Analysis | Date27/01/2025 | Catalog Number1093/E,207790050,207790250,207790010 |
- 1
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.