missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, 99.5%, for analysis, Thermo Scientific Chemicals

Sodium chloride, 99.5%, ClNa, CAS Number-7647-14-5, sodium chloride nacl, salt, common salt, rock salt, halite, purex, table salt, sodium chloride, saline, dendritis, 1kg, 99.4% min. (Argentometry), 10ppm max., 1461.0 deg.C, 50ppm max., 20ppm max., CHEBI:26710, White, 231-598-3, 13,280, 58.44

£21.51 - £228.14

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium;chloride
SMILES [Na+].[Cl-]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10092740
SDS View accessory products View Documents Promotion Details
Thermo Scientific Acros
207790010
1 kg Plastic Bottle
£21.51
1kg
Please to purchase this item. Need a web account? Register with us today!
10216410
SDS View Documents Promotion Details
Thermo Scientific Acros
207790050
5 kg Plastic Bottle
£63.44
5kg
Please to purchase this item. Need a web account? Register with us today!
10378573
SDS View Documents Promotion Details
Thermo Scientific Acros
207790250
25 kg Plastic Drum
£228.14
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-598-3

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium chloride, 99.5%, for analysis, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.