missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, pure, ca. 26% solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 387640010
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium chloride | |
Colorless | |
1 L | |
NaCl | |
15,8734 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium;chloride | |
5234 | |
58.44 | |
25 to 28% | |
1.202 |
7647-14-5, 7732-18-5 | |
Liquid | |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
Pure | |
Glass bottle | |
1.2020g/mL |
RUO – Research Use Only