missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, pure, ca. 26% solution in water, Thermo Scientific Chemicals
£24.00 - £47.10
Specifications
Chemical Name or Material | Sodium chloride |
---|---|
CAS | 7647-14-5, 7732-18-5 |
Color | Colorless |
Physical Form | Liquid |
Molecular Formula | ClNa |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10710945
|
Thermo Scientific Acros
387640010 |
1 L | Glass bottle |
£24.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10370244
|
Thermo Scientific Acros
387640025 |
2.5 L | Glass bottle |
£47.10
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium chloride | |
Colorless | |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
5234 | |
58.44 | |
25 to 28% | |
1.202 |
7647-14-5, 7732-18-5 | |
Liquid | |
NaCl | |
15,8734 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
58.44 | |
CHEBI:26710 | |
Pure | |
Glass bottle | |
1.2020g/mL |
Safety and Handling
missing translation for 'einecsNumber' : 231-598-3
RUO – Research Use Only