missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Product Code. 15693410 Shop All Honeywell Fluka Products6 x 500 g
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
25 kg
4 x 5 kg
6 x 1 kg
This item is not returnable.
View return policy
Product Code. 15693410
Brand: Honeywell-Fluka 314346X500G
This item is not returnable.
View return policy
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Especificaciones
Identificadores de productos químicos
CAS | 7647-14-5 |
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Especificaciones
Quantity | 6 x 500 g |
Linear Formula | NaCl |
UN Number | NONH for all modes of transport |
Solubility Information | Soluble in water, 100mg/mL |
IUPAC Name | sodium;chloride |
Formula Weight | 58.44 |
Percent Purity | ≥99.5% |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Sodium Chloride, Reag. Ph. Eur. reag. ISO |
Sugerencias de productos
You may be interested in these alternatives
-
Product Code. 10326390
-
Product Code. 10428420
-
Product Code. 15623410
-
Product Code. 15673410
-
Product Code. 10216410
-
-
Product Code. 15653410
-
Product Code. 15683410
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 7
-
Catalog No. 10428420
£72.05 / 1kg -
Catalog No. 10703811
£258.00 / 500g -
Catalog No. 10793611
£101.00 / 1kg -
Catalog No. 10117113
£25.71 / 100g -
Catalog No. 10112640
£50.40 / 1kg -
Catalog No. 10092740
£21.51 / 1kg -
Catalog No. 10401841
£22.81 / 500mL
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Sodium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™ >
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido