missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyclopentadienide, 2-3M in THF, Thermo Scientific Chemicals

Reagent for the preparation of metallocenes

Chemical Identifiers

CAS 4984-82-1
Molecular Formula C5H5Na
Molecular Weight (g/mol) 88.085
MDL Number MFCD00012161
InChI Key JQUZTGJSSQCTPV-UHFFFAOYSA-N
Synonym cyclopentadienylsodium, sodium cyclopenta-1,3-dien-1-ide, sodium cyclopentadienide, 2.0m solution in tetrahydrofuran
PubChem CID 21915903
IUPAC Name sodium;cyclopenta-1,3-diene
SMILES C1C=CC=[C-]1.[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326389
View Documents
Thermo Scientific Alfa Aesar
088300.JQ
0.1 mol N/A
N/A
11336389
View Documents
Thermo Scientific Alfa Aesar
088300.JT
0.5 mol N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4984-82-1
88.085
JQUZTGJSSQCTPV-UHFFFAOYSA-N
21915903
C1C=CC=[C-]1.[Na+]
C5H5Na
MFCD00012161
cyclopentadienylsodium, sodium cyclopenta-1,3-dien-1-ide, sodium cyclopentadienide, 2.0m solution in tetrahydrofuran
sodium;cyclopenta-1,3-diene

Specifications

4984-82-1
0.938 g/mL
Ether-like
MFCD00012161
Air and moisture sensitive
Reacts with water.
C1C=CC=[C-]1.[Na+]
88.085
88.09
Liquid
Pink to Red
−17°C (1°F)
C5H5Na
UN3399
cyclopentadienylsodium, sodium cyclopenta-1,3-dien-1-ide, sodium cyclopentadienide, 2.0m solution in tetrahydrofuran
JQUZTGJSSQCTPV-UHFFFAOYSA-N
sodium;cyclopenta-1,3-diene
21915903
2 to 3M in THF
Sodium cyclopentadienide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H250-H261-H302-H314-H335-H336-H351

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

missing translation for 'einecsNumber' : 225-636-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only