Learn More
Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
£32.66 - £243.21
Chemical Identifiers
CAS | 10049-21-5 |
---|---|
Molecular Formula | H2NaO4P x H2O |
Molecular Weight (g/mol) | 137.99 |
MDL Number | MFCD00149208 |
InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
Synonym | sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 |
PubChem CID | 516949 |
SMILES | O.[Na+].OP(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357327
|
Thermo Scientific Alfa Aesar
011591.30 |
250 g |
£32.66
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367327
|
Thermo Scientific Alfa Aesar
011591.A1 |
1 kg |
£82.31
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347327
|
Thermo Scientific Alfa Aesar
011591.A7 |
5 kg |
£243.21
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It is applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10049-21-5 | |
137.99 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
516949 |
H2NaO4P x H2O | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
O.[Na+].OP(O)(O)=O |
Specifications
100°C (in 2H2O) | |
Crystalline | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
sodium;dihydrogen phosphate;hydrate | |
516949 | |
98.0 to 102.0% | |
Odorless | |
Sodium dihydrogen phosphate monohydrate |
10049-21-5 | |
H2NaO4P x H2O | |
14,8660 | |
Highly soluble in water. Insoluble in ethanol,ether and chloroform. | |
O.[Na+].OP(O)(O)=O | |
137.99 | |
137.99 (119.98 Anhydrous) | |
ACS | |
Hygroscopic |
Safety and Handling
GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 231-449-2
missing translation for 'rtecsNumber' : WA1900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only